Assay Details
| Gene Symbol: JAZ3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_112667.4 | NP_566590.1 | 1 - 2 | 348 | 107 | |
| GenBank mRNA | AF372907.1 | - | 1 - 2 | 288 | 107 | |
| BT003044.1 | - | 1 - 2 | 87 | 107 | ||
More Information
Set Membership: |
|

JAZ3
Genomic Map