Assay Details
| Gene Symbol: JAZ3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001084705.3 | NP_001078174.1 | 2 - 3 | 1206 | 80 | |
| NM_001338310.1 | NP_001327107.1 | 2 - 3 | 854 | 80 | ||
| NM_112667.4 | NP_566590.1 | 2 - 3 | 541 | 80 | ||
| NM_202601.3 | NP_974330.1 | 1 - 2 | 1295 | 80 | ||
| GenBank mRNA | AF372907.1 | - | 2 - 3 | 481 | 80 | |
| AK222169.1 | - | 1 - 2 | 200 | 80 | ||
| BT003044.1 | - | 2 - 3 | 280 | 80 | ||
More Information
Set Membership: |
|

JAZ3
Genomic Map