Assay Details
| Gene Symbol: JAZ3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001084705.3 | NP_001078174.1 | 3 - 4 | 1426 | 66 | |
| NM_001338310.1 | NP_001327107.1 | 3 - 4 | 1074 | 66 | ||
| NM_112667.4 | NP_566590.1 | 3 - 4 | 761 | 66 | ||
| NM_202601.3 | NP_974330.1 | 2 - 3 | 1515 | 66 | ||
| GenBank mRNA | AF372907.1 | - | 3 - 4 | 701 | 66 | |
| AK222169.1 | - | 2 - 3 | 420 | 66 | ||
| BT003044.1 | - | 3 - 4 | 500 | 66 | ||
More Information
Set Membership: |
|

JAZ3
Genomic Map