Assay Details
| Gene Symbol: JAZ3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001084705.3 | NP_001078174.1 | 4 - 5 | 1519 | 71 | |
| NM_001338310.1 | NP_001327107.1 | 4 - 5 | 1167 | 71 | ||
| NM_112667.4 | NP_566590.1 | 4 - 5 | 854 | 71 | ||
| NM_202601.3 | NP_974330.1 | 3 - 4 | 1608 | 71 | ||
| GenBank mRNA | AF372907.1 | - | 4 - 5 | 794 | 71 | |
| AK222169.1 | - | 3 - 4 | 513 | 71 | ||
| BT003044.1 | - | 4 - 5 | 593 | 71 | ||
More Information
Set Membership: |
|

JAZ3
Genomic Map