Assay Details
| Gene Symbol: BGLU43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001084707.2 | NP_001078176.1 | 7 - 8 | 996 | 150 | |
| NM_001338322.1 | NP_001325994.1 | 10 - 11 | 1236 | 150 | ||
| NM_001338323.1 | NP_001325998.1 | 9 - 10 | 1195 | 150 | ||
| NM_001338326.1 | NP_001325996.1 | 6 - 7 | 764 | 150 | ||
| NM_112689.2 | NP_188435.2 | 10 - 11 | 1227 | 150 | ||
| GenBank mRNA | DQ446670.1 | - | 7 - 8 | 996 | 150 | |
More Information
Set Membership: |
|

BGLU43
Genomic Map