Assay Details
| Gene Symbol: NUC-L2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001338347.1 | NP_001325664.1 | 5 - 6 | 610 | 146 | |
| NM_001338348.1 | NP_001325663.1 | 6 - 7 | 643 | 146 | ||
| NM_112747.3 | NP_188491.1 | 7 - 8 | 675 | 146 | ||
| GenBank mRNA | DQ446672.1 | - | 7 - 8 | 666 | 146 | |
More Information
Set Membership: |
|

NUC-L2
Genomic Map