Assay Details
| Gene Symbol: HAF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001338377.1 | NP_001327580.1 | 10 - 11 | 2756 | 72 | |
| NM_112790.3 | NP_188534.2 | 9 - 10 | 2677 | 72 | ||
| GenBank mRNA | AY579213.1 | - | 9 - 10 | 2677 | 72 | |
More Information
Set Membership: |
|

HAF2
Genomic Map