Assay Details
| Gene Symbol: MGT4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_112854.4 | NP_188598.2 | 1 - 2 | 476 | 84 | |
| GenBank mRNA | AY150288.1 | - | 1 - 2 | 452 | 84 | |
| BT000458.1 | - | 1 - 2 | 472 | 84 | ||
| BT006622.1 | - | 1 - 2 | 433 | 84 | ||
More Information
Set Membership: |
|

MGT4
Genomic Map