Assay Details
| Gene Symbol: MGT4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_112854.4 | NP_188598.2 | 3 - 4 | 819 | 76 | |
| GenBank mRNA | BT000458.1 | - | 3 - 4 | 815 | 76 | |
| BT006622.1 | - | 3 - 4 | 776 | 76 | ||
More Information
Set Membership: |
|

MGT4
Genomic Map