Assay Details
| Gene Symbol: ATM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001161156.2 | NP_001154628.2 | 2 - 3 | 669 | 80 | |
| NM_001338431.1 | NP_001325652.1 | 2 - 3 | 669 | 80 | ||
| NM_001338432.1 | NP_001325654.1 | 2 - 3 | 631 | 80 | ||
| NM_001338433.1 | NP_001325653.1 | 2 - 3 | 513 | 80 | ||
| NM_112886.4 | NP_188630.1 | 2 - 3 | 669 | 80 | ||
| GenBank mRNA | BT001941.1 | - | 1 - 2 | 336 | 80 | |
More Information
Set Membership: |
|

ATM1
Genomic Map