Assay Details
| Gene Symbol: TTN9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001338449.1 | NP_001327503.1 | 3 - 4 | 334 | 126 | |
| NM_112898.4 | NP_188642.2 | 2 - 3 | 432 | 126 | ||
| NM_202613.3 | NP_974342.1 | 3 - 4 | 362 | 126 | ||
| GenBank mRNA | AX765848.1 | - | 2 - 3 | 116 | 126 | |
| AY063778.1 | - | 2 - 3 | 412 | 126 | ||
| AY123015.1 | - | 2 - 3 | 116 | 126 | ||
More Information
Set Membership: |
|

TTN9
Genomic Map