Assay Details
| Gene Symbol: TTN9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001338448.1 | NP_001327502.1 | 3 - 4 | 698 | 89 | |
| NM_001338449.1 | NP_001327503.1 | 5 - 6 | 494 | 89 | ||
| NM_112898.4 | NP_188642.2 | 4 - 5 | 592 | 89 | ||
| NM_202613.3 | NP_974342.1 | 5 - 6 | 522 | 89 | ||
| GenBank mRNA | AX765848.1 | - | 4 - 5 | 276 | 89 | |
| AY063778.1 | - | 4 - 5 | 572 | 89 | ||
| AY123015.1 | - | 4 - 5 | 276 | 89 | ||
More Information
Set Membership: |
|

TTN9
Genomic Map