Assay Details
| Gene Symbol: HMGB5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001342361.1 | NP_001329802.1 | 2 - 3 | 242 | 67 | |
| NM_119722.3 | NP_195282.1 | 3 - 4 | 233 | 67 | ||
| GenBank mRNA | AK317173.1 | - | 3 - 4 | 201 | 67 | |
| AY085615.1 | - | 3 - 4 | 205 | 67 | ||
| BT025597.1 | - | 2 - 3 | 140 | 67 | ||
| Y14075.1 | - | 140 | 67 | |||
More Information
Set Membership: |
|

HMGB5
Genomic Map