Assay Details
| Gene Symbol: NTL9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001125650.1 | NP_001119122.1 | 4 - 5 | 1353 | 64 | |
| NM_001203998.1 | NP_001190927.1 | 4 - 5 | 1429 | 64 | ||
| NM_119723.5 | NP_567986.3 | 4 - 5 | 1427 | 64 | ||
More Information
Set Membership: |
|

NTL9
Genomic Map