Assay Details
| Gene Symbol: NTL9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_119723.5 | NP_567986.3 | 5 - 6 | 1552 | 71 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

NTL9| Gene Symbol: NTL9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_119723.5 | NP_567986.3 | 5 - 6 | 1552 | 71 | |
Set Membership: |
|