Assay Details
| Gene Symbol: CRH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_119796.4 | NP_195351.2 | 11 - 12 | 1223 | 118 | |
| GenBank mRNA | BT020539.1 | - | 11 - 12 | 1134 | 118 | |
More Information
Set Membership: |
|

CRH1
Genomic Map