Assay Details
| Gene Symbol: CRH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_119796.4 | NP_195351.2 | 14 - 15 | 1481 | 98 | |
| GenBank mRNA | BT020539.1 | - | 14 - 15 | 1392 | 98 | |
More Information
Set Membership: |
|

CRH1
Genomic Map