Assay Details
| Gene Symbol: CRH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_119796.4 | NP_195351.2 | 1 - 2 | 311 | 65 | |
| GenBank mRNA | BT020539.1 | - | 1 - 2 | 222 | 65 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CRH1| Gene Symbol: CRH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_119796.4 | NP_195351.2 | 1 - 2 | 311 | 65 | |
| GenBank mRNA | BT020539.1 | - | 1 - 2 | 222 | 65 | |
Set Membership: |
|