Assay Details
| Gene Symbol: CRT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_119797.4 | NP_568000.1 | 13 - 14 | 1473 | 64 | |
| GenBank mRNA | BT001940.1 | - | 13 - 14 | 1299 | 64 | |
More Information
Set Membership: |
|

CRT1
Genomic Map