Assay Details
| Gene Symbol: CRT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_119797.4 | NP_568000.1 | 1 - 2 | 414 | 95 | |
| GenBank mRNA | BT001940.1 | - | 1 - 2 | 240 | 95 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CRT1| Gene Symbol: CRT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_119797.4 | NP_568000.1 | 1 - 2 | 414 | 95 | |
| GenBank mRNA | BT001940.1 | - | 1 - 2 | 240 | 95 | |
Set Membership: |
|