Assay Details
| Gene Symbol: LCB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036719.2 | NP_001031796.1 | 5 - 6 | 592 | 91 | |
| NM_001342410.1 | NP_001329288.1 | 5 - 6 | 862 | 91 | ||
| NM_119811.3 | NP_568005.1 | 5 - 6 | 862 | 91 | ||
| GenBank mRNA | AB063254.1 | - | 4 - 5 | 404 | 91 | |
| AY120759.1 | - | 5 - 6 | 680 | 91 | ||
| BT000131.1 | - | 4 - 5 | 383 | 91 | ||
More Information
Set Membership: |
|

LCB1
Genomic Map