Assay Details
| Gene Symbol: LCB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036719.2 | NP_001031796.1 | 9 - 10 | 1115 | 64 | |
| NM_001342410.1 | NP_001329288.1 | 9 - 10 | 1385 | 64 | ||
| NM_119811.3 | NP_568005.1 | 9 - 10 | 1385 | 64 | ||
| GenBank mRNA | AB063254.1 | - | 8 - 9 | 927 | 64 | |
| AY120759.1 | - | 9 - 10 | 1203 | 64 | ||
| BT000131.1 | - | 8 - 9 | 906 | 64 | ||
More Information
Set Membership: |
|

LCB1
Genomic Map