Assay Details
| Gene Symbol: NPY5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_119923.4 | NP_568030.1 | 4 - 5 | 2418 | 67 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

NPY5| Gene Symbol: NPY5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_119923.4 | NP_568030.1 | 4 - 5 | 2418 | 67 | |
Set Membership: |
|