Assay Details
| Gene Symbol: ETC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001331271.1 | NP_001321146.1 | 1 - 2 | 293 | 93 | |
| NM_001331272.1 | NP_001321147.1 | 1 - 2 | 183 | 93 | ||
| NM_100020.4 | NP_171645.1 | 1 - 2 | 293 | 93 | ||
| GenBank mRNA | AY519518.1 | - | 1 - 2 | 69 | 93 | |
More Information
Set Membership: |
|

ETC1
Genomic Map