Assay Details
| Gene Symbol: AGL28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_100035.3 | NP_171660.1 | 1 - 2 | 319 | 79 | |
| GenBank mRNA | AB493420.1 | - | 1 - 2 | 311 | 79 | |
| DQ446218.1 | - | 1 - 2 | 311 | 79 | ||
More Information
Set Membership: |
|

AGL28
Genomic Map