Assay Details
| Gene Symbol: SPPL4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035853.1 | NP_001030930.1 | 9 - 10 | 1470 | 58 | |
| NM_001331286.1 | NP_001318903.1 | 9 - 10 | 1750 | 58 | ||
| NM_001331287.1 | NP_001323100.1 | 9 - 10 | 1754 | 58 | ||
| NM_100047.4 | NP_171671.2 | 10 - 11 | 1478 | 58 | ||
| GenBank mRNA | AK229822.1 | - | 10 - 11 | 1327 | 58 | |
More Information
Set Membership: |
|

SPPL4
Genomic Map