Assay Details
| Gene Symbol: SPPL4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035853.1 | NP_001030930.1 | 4 - 5 | 933 | 75 | |
| NM_001331286.1 | NP_001318903.1 | 4 - 5 | 1213 | 75 | ||
| NM_001331287.1 | NP_001323100.1 | 4 - 5 | 1217 | 75 | ||
| NM_100047.4 | NP_171671.2 | 5 - 6 | 941 | 75 | ||
| GenBank mRNA | AK229822.1 | - | 5 - 6 | 790 | 75 | |
More Information
Set Membership: |
|

SPPL4
Genomic Map