Assay Details
| Gene Symbol: AT1G01980 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_100078.3 | NP_171700.1 | 1 | 1307 | 92 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AT1G01980| Gene Symbol: AT1G01980 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_100078.3 | NP_171700.1 | 1 | 1307 | 92 | |
Set Membership: |
|