Assay Details
| Gene Symbol: FAF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_100199.2 | NP_563678.1 | 1 | 1797 | 85 | |
| GenBank mRNA | AK118027.1 | - | 1 | 541 | 85 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

FAF2| Gene Symbol: FAF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_100199.2 | NP_563678.1 | 1 | 1797 | 85 | |
| GenBank mRNA | AK118027.1 | - | 1 | 541 | 85 | |
Set Membership: |
|