Assay Details
| Gene Symbol: POR C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_100243.4 | NP_171860.1 | 2 - 3 | 339 | 67 | |
| GenBank mRNA | AB035746.1 | - | 2 - 3 | 187 | 67 | |
| AK226626.1 | - | 210 | 67 | |||
| AY048263.1 | - | 2 - 3 | 205 | 67 | ||
| AY088529.1 | - | 2 - 3 | 211 | 67 | ||
| AY133569.1 | - | 2 - 3 | 163 | 67 | ||
More Information
Set Membership: |
|

POR C
Genomic Map