Assay Details
| Gene Symbol: POR C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035871.1 | NP_001030948.1 | 3 - 4 | 739 | 137 | |
| NM_100243.4 | NP_171860.1 | 3 - 4 | 873 | 137 | ||
| GenBank mRNA | AB035746.1 | - | 3 - 4 | 721 | 137 | |
| AK226626.1 | - | 744 | 137 | |||
| AY048263.1 | - | 3 - 4 | 739 | 137 | ||
| AY088529.1 | - | 3 - 4 | 745 | 137 | ||
| AY133569.1 | - | 3 - 4 | 697 | 137 | ||
More Information
Set Membership: |
|

POR C
Genomic Map