Assay Details
| Gene Symbol: ADK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_100274.3 | NP_563695.2 | 5 - 6 | 1175 | 145 | |
| GenBank mRNA | AY065409.1 | - | 5 - 6 | 764 | 145 | |
| AY075642.1 | - | 5 - 6 | 1093 | 145 | ||
| AY096519.1 | - | 5 - 6 | 424 | 145 | ||
| AY101520.1 | - | 5 - 6 | 424 | 145 | ||
| AY943848.1 | - | 5 - 6 | 424 | 145 | ||
More Information
Set Membership: |
|

ADK1
Genomic Map