Assay Details
| Gene Symbol: SUVR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001331458.1 | NP_001320725.1 | 6 - 7 | 947 | 89 | |
| NM_001331459.1 | NP_001320724.1 | 4 - 5 | 568 | 89 | ||
| NM_100286.4 | NP_171901.3 | 5 - 6 | 968 | 89 | ||
| GenBank mRNA | AF394239.1 | - | 3 - 4 | 435 | 89 | |
More Information
Set Membership: |
|

SUVR1
Genomic Map