Assay Details
| Gene Symbol: SUVR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001331458.1 | NP_001320725.1 | 11 - 12 | 2435 | 88 | |
| NM_001331459.1 | NP_001320724.1 | 9 - 10 | 2056 | 88 | ||
| NM_100286.4 | NP_171901.3 | 10 - 11 | 2456 | 88 | ||
| GenBank mRNA | AF394239.1 | - | 8 - 9 | 1923 | 88 | |
More Information
Set Membership: |
|

SUVR1
Genomic Map