Assay Details
| Gene Symbol: AT1G04810 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_100359.3 | NP_171973.1 | 12 - 13 | 2878 | 69 | |
| GenBank mRNA | BT004007.1 | - | 12 - 13 | 2778 | 69 | |
More Information
Set Membership: |
|

AT1G04810
Genomic Map