Assay Details
| Gene Symbol: PDIL2-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_100376.5 | NP_171990.3 | 7 - 8 | 1103 | 121 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PDIL2-2| Gene Symbol: PDIL2-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_100376.5 | NP_171990.3 | 7 - 8 | 1103 | 121 | |
Set Membership: |
|