Assay Details
| Gene Symbol: SPPL5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001123763.1 | NP_001117235.1 | 2 - 3 | 388 | 72 | |
| NM_001331585.1 | NP_001323185.1 | 2 - 3 | 529 | 72 | ||
| NM_100463.3 | NP_172073.2 | 2 - 3 | 592 | 72 | ||
| GenBank mRNA | AB330675.1 | - | 2 - 3 | 294 | 72 | |
More Information
Set Membership: |
|

SPPL5
Genomic Map