Assay Details
| Gene Symbol: CAT9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001331601.1 | NP_001322444.1 | 2 - 3 | 986 | 113 | |
| NM_001331602.1 | NP_001322445.1 | 3 - 4 | 888 | 113 | ||
| NM_001331603.1 | NP_001322446.1 | 3 - 4 | 891 | 113 | ||
| NM_100475.4 | NP_563754.1 | 3 - 4 | 901 | 113 | ||
| GenBank mRNA | AF360321.1 | - | 3 - 4 | 877 | 113 | |
| AY113890.1 | - | 3 - 4 | 843 | 113 | ||
More Information
Set Membership: |
|

CAT9
Genomic Map