Assay Details
| Gene Symbol: CAT9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001331601.1 | NP_001322444.1 | 4 - 5 | 1385 | 98 | |
| NM_001331602.1 | NP_001322445.1 | 5 - 6 | 1287 | 98 | ||
| NM_001331603.1 | NP_001322446.1 | 5 - 6 | 1290 | 98 | ||
| NM_100475.4 | NP_563754.1 | 5 - 6 | 1300 | 98 | ||
More Information
Set Membership: |
|

CAT9
Genomic Map