Assay Details
| Gene Symbol: NIH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001331643.1 | NP_001321998.1 | 6 - 7 | 972 | 85 | |
| NM_100544.3 | NP_172152.1 | 6 - 7 | 995 | 85 | ||
More Information
Set Membership: |
|

NIH
Genomic Map