Assay Details
| Gene Symbol: AT1G06690 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_100546.4 | NP_563770.1 | 3 - 4 | 506 | 69 | |
| GenBank mRNA | AY050325.1 | - | 3 - 4 | 400 | 69 | |
| BT001002.1 | - | 3 - 4 | 375 | 69 | ||
More Information
Set Membership: |
|

AT1G06690
Genomic Map