Assay Details
| Gene Symbol: AT1G06690 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_100546.4 | NP_563770.1 | 7 - 8 | 934 | 96 | |
| GenBank mRNA | AY050325.1 | - | 7 - 8 | 828 | 96 | |
| BT001002.1 | - | 7 - 8 | 803 | 96 | ||
More Information
Set Membership: |
|

AT1G06690
Genomic Map