Assay Details
| Gene Symbol: DFO | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001331678.1 | NP_001322524.1 | 2 - 3 | 355 | 66 | |
| NM_001331679.1 | NP_001322523.1 | 2 - 3 | 355 | 66 | ||
| NM_001331680.1 | NP_001322522.1 | 1 - 2 | 300 | 66 | ||
| NM_100580.2 | NP_172187.1 | 2 - 3 | 355 | 66 | ||
| GenBank mRNA | AY090924.1 | - | 2 - 3 | 355 | 66 | |
More Information
Set Membership: |
|

DFO
Genomic Map