Assay Details
| Gene Symbol: PCNA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_100611.4 | NP_172217.1 | 1 - 2 | 488 | 98 | |
| GenBank mRNA | AF462821.1 | - | 1 - 2 | 371 | 98 | |
| AY086852.1 | - | 1 - 2 | 383 | 98 | ||
| AY098969.1 | - | 1 - 2 | 321 | 98 | ||
More Information
Set Membership: |
|

PCNA1
Genomic Map