Assay Details
| Gene Symbol: AT3G20760 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_112967.4 | NP_188712.4 | 3 - 4 | 988 | 139 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AT3G20760| Gene Symbol: AT3G20760 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_112967.4 | NP_188712.4 | 3 - 4 | 988 | 139 | |
Set Membership: |
|