Assay Details
| Gene Symbol: CHL-CPN10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_130029.4 | NP_566022.1 | 4 - 5 | 863 | 66 | |
| GenBank mRNA | AB051163.1 | - | 4 - 5 | 316 | 66 | |
More Information
Set Membership: |
|

CHL-CPN10
Genomic Map