Assay Details
| Gene Symbol: AT2G45290 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001337121.1 | NP_001324258.1 | 6 - 7 | 1435 | 68 | |
| NM_130092.3 | NP_566041.2 | 6 - 7 | 1501 | 68 | ||
More Information
Set Membership: |
|

AT2G45290
Genomic Map