Assay Details
| Gene Symbol: SLD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_130183.4 | NP_182144.1 | 1 | 1558 | 86 | |
| Gene Symbol: AT2G46200 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001337179.1 | NP_001324669.1 | 1 | 161 | 86 | |
| NM_180117.2 | NP_850448.1 | 1 | 161 | 86 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001337179.1, NM_180117.2 transcripts.
Set Membership: |
|

SLD2
Genomic Map