Assay Details
| Gene Symbol: RS31a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_130226.6 | NP_182184.1 | 5 - 6 | 869 | 66 | |
| NM_201973.6 | NP_973702.1 | 5 - 6 | 1388 | 66 | ||
| GenBank mRNA | AK317202.1 | - | 5 - 6 | 870 | 66 | |
| AY065237.1 | - | 5 - 6 | 821 | 66 | ||
| BT000995.1 | - | 4 - 5 | 725 | 66 | ||
More Information
Set Membership: |
|

RS31a
Genomic Map