Assay Details
| Gene Symbol: AIP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_100752.4 | NP_563827.1 | 2 - 3 | 319 | 68 | |
| GenBank mRNA | AK227759.1 | - | 2 - 3 | 195 | 68 | |
| BT004788.1 | - | 2 - 3 | 132 | 68 | ||
More Information
Set Membership: |
|

AIP3
Genomic Map